Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ascorbic Acid, Crystalline Powder, FCC, 99-100.5%, Spectrum™

Click to view available options
Quantity:
25 g
100 g
500 g
2.5 kg
12 kg
50 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
50-81-7
C6H8O6
176.12
TYQCGQRIZGCHNB-UHFFFAOYNA-N
2-(1,2-dihydroxyethyl)-4,5-dihydroxy-2,3-dihydrofuran-3-one
OCC(O)C1OC(O)=C(O)C1=O
50-81-7
C6H8O6
176.12
TYQCGQRIZGCHNB-UHFFFAOYNA-N
2-(1,2-dihydroxyethyl)-4,5-dihydroxy-2,3-dihydrofuran-3-one
OCC(O)C1OC(O)=C(O)C1=O

A1371, 50-81-7, C6H8O6

Spectrum™ Ascorbic Acid, Crystalline Powder, FCC is commonly used as antioxidant food additive. Spectrum Chemical offers over 300 Food Grade (FCC) chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Packaging Fiber Drum
Specific Rotation +20.5° to +21.5°
Ignition Residue 0.001
Quantity 50 kg
CAS Max % 1
Percent Purity 99 to 100.5%