Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ascorbic Acid, Crystalline Powder, USP, 99-100.5%, Spectrum™

Click to view available options
Quantity:
25 g
100 g
500 g
1 kg
5 kg
12 kg
50 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
50-81-7
C6H8O6
176.12
TYQCGQRIZGCHNB-UHFFFAOYNA-N
2-(1,2-dihydroxyethyl)-4,5-dihydroxy-2,3-dihydrofuran-3-one
OCC(O)C1OC(O)=C(O)C1=O
50-81-7
C6H8O6
176.12
TYQCGQRIZGCHNB-UHFFFAOYNA-N
2-(1,2-dihydroxyethyl)-4,5-dihydroxy-2,3-dihydrofuran-3-one
OCC(O)C1OC(O)=C(O)C1=O

AS105, 50-81-7, C6H8O6

Spectrum™ Ascorbic Acid, Crystalline Powder, USP , Vitamin C, is a dietary supplement used to treat and prevent vitamin C deficiency. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Ignition Residue 0.1%
Packaging Amber Glass Bottle
Specific Rotation +20.5° to +21.5°
Quantity 1 kg
CAS Max % 100%
Percent Purity 99 to 100.5%