Learn More
BioVision IncSupplier Diversity Partner Ascorbic Acid Colorimetric/Fluorometric Assay Kit: Colorimetric & Fluorometric Assay for detecting Ascorbic Acid in various Biological Samples such as Serum etc. within 5 min. Convenient, Accurate & Reliable. Detection Range: 0.01-10 nmol. DFS Item
Manufacturer:  BioVision IncSupplier Diversity Partner K661100
Ascorbic Acid (Vitamin C) plays an important role in many biological processes. It is a potent anti-oxidant, anti-inflammatory, anti-viral agent, and an immune stimulant and is present is a wide variety of foods and biological specimens. It is important to be able to monitor ascorbic acid content in these different samples. BioVision’s Ascorbic Acid Assay Kit provides a rapid, simple, and sensitive means of detecting ascorbic acid in various biological samples. In this assay, our proprietary catalyst oxidizes ascorbic acid to produce a product that interacts with the ascorbic acid probe, generating color and fluorescence. Ascorbic acid can be easily determined by either colorimetric (spectrophotometry at λ = 570 nm) or fluorometric (Ex/Em = 535/587 nm) methods. The assay can detect 0.01-10 nmol of ascorbic acid per assay in various samples. BioVision charges an Ice pack and Insulated Container fee of $25.00 for each order.
Catalog No. 50-111-9822