Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ascorbic Acid, Reagent, ACS, 99%, Spectrum™

Click to view available options
Quantity:
100 g
500 g
5 kg
12 kg
Packaging:
Amber Glass Bottle
Poly Pail
50-81-7
C6H8O6
176.12
TYQCGQRIZGCHNB-UHFFFAOYNA-N
2-(1,2-dihydroxyethyl)-4,5-dihydroxy-2,3-dihydrofuran-3-one
OCC(O)C1OC(O)=C(O)C1=O
50-81-7
C6H8O6
176.12
TYQCGQRIZGCHNB-UHFFFAOYNA-N
2-(1,2-dihydroxyethyl)-4,5-dihydroxy-2,3-dihydrofuran-3-one
OCC(O)C1OC(O)=C(O)C1=O

A1370, 50-81-7, C6H8O6

Spectrum™ Ascorbic Acid, Reagent, ACS , is an organic compound that occurs naturally and has antioxidant properties. It is a form of vitamin C and is classed as a reductone and used as an antioxidant food additive. Other uses include reactant in photographic developer solutions, antioxidant in fluorescence microscopy to increase fluorescent signals and chemically retard dye photobleaching. As an ACS grade Reagent, Spectrum Chemical manufactured Ascorbic Acid is used as the quality standard against which other substances are graded and has met the toughest regulatory standards for quality and pureness.
Packaging Poly Pail
Quantity 12 kg
Specific Rotation +20.5° to +21.5°
CAS Max % 100%
Percent Purity 99%