Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ascorbic Acid, USP, 99-100.5%, Spectrum™

Click to view available options
Quantity:
25 g
100 g
500 g
1 kg
5 kg
12 kg
50 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
50-81-7
C6H8O6
176.12
TYQCGQRIZGCHNB-UHFFFAOYNA-N
2-(1,2-dihydroxyethyl)-4,5-dihydroxy-2,3-dihydrofuran-3-one
OCC(O)C1OC(O)=C(O)C1=O
50-81-7
C6H8O6
176.12
TYQCGQRIZGCHNB-UHFFFAOYNA-N
2-(1,2-dihydroxyethyl)-4,5-dihydroxy-2,3-dihydrofuran-3-one
OCC(O)C1OC(O)=C(O)C1=O

AS102, 50-81-7, C6H8O6

Spectrum™ Ascorbic Acid, USP , Vitamin C, is a dietary supplement used to treat and prevent vitamin C deficiency. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Packaging Poly Pail
Specific Rotation +20.5° to +21.5°
Quantity 12 kg
Ignition Residue 0.001
CAS Max % 1
Percent Purity 99 to 100.5%