Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ascorbyl Palmitate, FCC, 95%, Spectrum™

Click to view available options
Quantity:
25 g
125 g
500 g
5 kg
Packaging:
Poly Bottle
Poly Pail
137-66-6
C22H38O7
414.54
NULLRXHRYOXSEW-UHFFFAOYNA-N
2-(4,5-dihydroxy-3-oxo-2,3-dihydrofuran-2-yl)-2-hydroxyethyl hexadecanoate
CCCCCCCCCCCCCCCC(=O)OCC(O)C1OC(O)=C(O)C1=O
137-66-6
C22H38O7
414.54
NULLRXHRYOXSEW-UHFFFAOYNA-N
2-(4,5-dihydroxy-3-oxo-2,3-dihydrofuran-2-yl)-2-hydroxyethyl hexadecanoate
CCCCCCCCCCCCCCCC(=O)OCC(O)C1OC(O)=C(O)C1=O

A1374, 137-66-6, C22H38O7

Spectrum™ Ascorbyl Palmitate, FCC is used as an antioxidant food additive (E number E304). The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Packaging Poly Bottle
Ignition Residue 0.001
Specific Rotation +21° to +24°
Loss on Drying 0.02
Quantity 125 g
Melting Range 107-117°C
CAS Max % 1
Percent Purity 95%