Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ascorbyl Palmitate, NF, 95-100.5%, Spectrum™

Click to view available options
Quantity:
25 g
125 g
500 g
5 kg
25 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
137-66-6
C22H38O7
414.54
NULLRXHRYOXSEW-UHFFFAOYNA-N
2-(4,5-dihydroxy-3-oxo-2,3-dihydrofuran-2-yl)-2-hydroxyethyl hexadecanoate
CCCCCCCCCCCCCCCC(=O)OCC(O)C1OC(O)=C(O)C1=O
137-66-6
C22H38O7
414.54
NULLRXHRYOXSEW-UHFFFAOYNA-N
2-(4,5-dihydroxy-3-oxo-2,3-dihydrofuran-2-yl)-2-hydroxyethyl hexadecanoate
CCCCCCCCCCCCCCCC(=O)OCC(O)C1OC(O)=C(O)C1=O

A1372, 137-66-6, C22H38O7

Spectrum™ Ascorbyl Palmitate, NF is often used to promote flow of tableting powders and to prevent their sticking in tablet presses. Spectrum Chemical NF products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Ignition Residue 0.1%
Loss on Drying 2%
Packaging Poly Pail
Quantity 500 g
Specific Rotation +21° to +24°
CAS Max % 100%
Percent Purity 95 to 100.5%