Aseptic Control Products™ VioNex™ Antiseptic Gel

Waterless gel for hand cleaning when soap and water are not available

Manufacturer: ASEPTIC CONTROL PRODUCTS 101624

 View more versions of this product

Catalog No. 50043456B

  Check Availability
Add to Cart

    Specifications

    Size (English) 4 oz.
    Size (Metric) 118mL
    Quantity 24 Case
    Certifications/Compliance OSHA
    Composition PCMX and Nonoxynol-9
    For Use With (Application) Effective against range of micoorganisms including TB, Staph and E. coli
    Format Gel
    Product Type VioNex™ Antiseptic Hand Gel

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit