Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ASF1a Mouse anti-Human, Clone: 2A5F8, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP261683

Catalog No. NBP2-61683


Add to cart

Description

Description

ASF1a Monoclonal antibody specifically detects ASF1a in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA.
Specifications

Specifications

ASF1a
2A5F8
Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, ELISA 1:100-1:2000
Purified
ASF1A
Purified recombinant fragment of human ASF1A (AA: 1-204) expressed in E. Coli.
23 kDa
0.1 ml
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Monoclonal
Human
ELISA, Flow Cytometry, Western Blot
Unconjugated
PBS with 0.05% Sodium Azide
ASF1 silencing function 1 homolog A (S. cerevisiae), CCG1-interacting factor A, CIACGI-98, DKFZP547E2110, hAsf1, hAsf1a, hCIA, histone chaperone ASF1A, HSPC146, silencing function 1A, silencing function protein 1 homolog A
Mouse
IgG1
Protein G purified
DNA Repair
Primary
25842
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit