Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ASF1A Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Thermo Scientific PA599078

Catalog No. PIPA599078


Add to cart

Description

Description

This gene encodes a member of the H3/H4 family of histone chaperone proteins and is similar to the anti-silencing function-1 gene in yeast. The protein is a key component of a histone donor complex that functions in nucleosome assembly. It interacts with histones H3 and H4, and functions together with a chromatin assembly factor during DNA replication and repair.
Specifications

Specifications

ASF1A
Polyclonal
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q9Y294
ASF1A
Recombit Human Histone chaperone ASF1A protein (1-204AA)
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
ASF1A
Liquid
Rabbit
IgG
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
25842
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit