Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ASF1b Mouse anti-Human, Alexa Fluor™ 350, Clone: 1D4, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer: Novus Biologicals NBP242676AF350
Description
ASF1b Monoclonal specifically detects ASF1b in Human samples. It is validated for Western Blot, ELISA.Specifications
ASF1b | |
1D4 | |
Western Blot, ELISA | |
Purified, Conjugated | |
ASF1B | |
Recombinant human ASF1B (1-202aa) purified from E. coli | |
Protein A purified | |
Store at 4°C in the dark. | |
Monoclonal | |
Human |
Western Blot, ELISA | |
Alexa Fluor 350 | |
50 mM sodium borate with 0.05% sodium azide | |
ASF1 silencing function 1 homolog B (S. cerevisiae), CCG1-interacting factor A-II, CIA-II, FLJ10604, hAsf1, hAsf1b, hCIA-II, histone chaperone ASF1B, silencing function protein 1 homolog B | |
Mouse | |
IgG2b Kappa | |
100 μL | |
Primary | |
55723.0 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title