Learn More
ASF1B Rabbit anti-Human, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 222581AP
Catalog No. 50-173-3801
Add to cart
Description
Description
This antibody specifically reacts with the 19 kDa ASF1B protein.
This gene encodes a member of the H3/H4 family of histone chaperone proteins and is similar to the anti-silencing function-1 gene in yeast. The encoded protein is the substrate of the tousled-like kinase family of cell cycle-regulated kinases, and may play a key role in modulating the nucleosome structure of chromatin by ensuring a constant supply of histones at sites of nucleosome assembly.
Specifications
Specifications
ASF1B
0.2 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q9NVP2
ASF1B, CCG1 interacting factor A II, CIA II, hAsf1, hAsf1b, hCIA II, Histone chaperone ASF1B
Rabbit
IgG
150 μL
-20°C
Primary
55723
Immunocytochemistry, Immunofluorescence, Western Blot
Unconjugated
ASF1B
Liquid
ASF1B
ASF1B Fusion Protein Ag17763
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human