Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ASGPR1 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
$152.22 - $436.00
Specifications
Antigen | ASGPR1 |
---|---|
Applications | Western Blot, Immunohistochemistry |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP15438520
|
Novus Biologicals
NBP15438520UL |
20 μL |
Each for $152.22
|
|
NBP154385
|
Novus Biologicals
NBP154385 |
100 μL |
Each for $436.00
|
|
Description
ASGPR1 Polyclonal specifically detects ASGPR1 in Human samples. It is validated for Western Blot.Specifications
ASGPR1 | |
Polyclonal | |
Purified | |
RUO | |
Q6FGQ5 | |
432 | |
IgG | |
Protein A purified | |
33 kDa |
Western Blot, Immunohistochemistry | |
Unconjugated | |
Rabbit | |
PBS and 2% Sucrose with 0.09% Sodium Azide | |
ASGPR, ASGPR 1, ASGP-R 1, asialoglycoprotein receptor 1, CLEC4H1ASGPR1, C-type lectin domain family 4 member H1, Hepatic lectin H1, HL-1 | |
ASGR1 | |
Primary | |
Store at -20C. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title