Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ASGR2 Mouse anti-Human, Clone: OTI2A12, liquid, TrueMAB
Mouse Monoclonal Antibody
Manufacturer: OriGene TA505099S
Description
ASGR2 is a cell surface receptor that binds to galactose-terminated glycoproteins. It transports these glycoproteins via a series of membrane vesicles and tubules to an acidic-sorting organelle where the receptor and ligand dissociates. Then the receptor is recycled back to the cell surface. There are four alternatively spliced transcript variants of this gene. This gene has multiple polyadenylation sites.Specifications
ASGR2 | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
P07307 | |
ASGP-R2, ASGPR2, CLEC4H2, HBXBP, HL-2 | |
Mouse | |
IgG2b | |
30 μL | |
-20° C, Avoid Freeze/Thaw Cycles | |
Primary | |
433 |
Immunohistochemistry (Paraffin), Western Blot | |
OTI2A12 | |
Unconjugated | |
ASGR2 | |
Liquid | |
ASGR2 | |
Full length human recombit protein of human ASGR2 produced in HEK293T cell. | |
Affinity Chromatography | |
RUO | |
Antibody | |
Monoclonal | |
Human |
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title