Learn More
ASH2L Rabbit anti-Mouse, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP259292
Catalog No. NBP259292
Add to cart
Description
Description
ASH2L Polyclonal antibody specifically detects ASH2L in Mouse samples. It is validated for Western Blot, ELISA, Immunofluorescence.
Specifications
Specifications
ASH2L
Unconjugated
Serum with 0.01% Sodium Azide
ASH2, ash2 (absent, small, or homeotic)-like (Drosophila), ASH2L1ash2 (absent, small, or homeotic, Drosophila, homolog)-like, ASH2L2ASH2-like protein, Bre2, set1/Ash2 histone methyltransferase complex subunit ASH2
Rabbit
IgG
100 ul
DNA Repair, Transcription Factors and Regulators
Primary
9070
Western Blot, ELISA, Immunofluorescence
Western Blot 1:500-1:1000, ELISA 1:100 - 1:500, Immunofluorescence 1:200
Antiserum
ASH2L
The exact immunogen is propietary information.
Whole Antiserum
RUO
Store at -20C short term. Aliquot and store at -80C long term. Avoid freeze-thaw cycles.
Polyclonal
Mouse