Learn More
AGI NORTH AMERICA INC Syrris Asia Flow Starter System for chemists eager to begin using flow chemistry - the system comes with everything you need
SDP
Supplier:  AGI NORTH AMERICA INC 40002AFBF
Encompass_Preferred
The easy-to-use and affordable Syrris Asia Flow Starter system contains all the essential flow system parts required for you to begin your flow chemistry journey.The Asia flow system’s unique modular design is completely future-proof and can be seamlessly upgraded with additional features and modules as the user experience with flow chemistry grows.Included:-Asia Syringe Pump with two independent flow channels each with an integrated pressure sensor and accurate flow rates of 50uL to 10mL/min-Asia Chip Climate Controller with accurate temperature control of -15 deg C to 150 deg C-Asia 1 mL Reactor Chip with excellent chemical compatibility-Asia Pressure Controller with pressure control up to 20bar in 0.1bar resolution-Asia Flow Starter System Tubing Kit to connect your flow systemOrder informationThe Asia Syringe Pump can be supplied with alternative syringe sizes for precise and accurate control of lower flow rates.
Catalog No. NC2417597
Only null left
Add to Cart
Request a Quote
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.