Learn More
Asialoglycoprotein Receptor 2 Rabbit anti-Human, Alexa Fluor 350, Clone: 2327C, R&D Systems
Rabbit Monoclonal Antibody
Manufacturer:  R&D Systems FAB9970U100UG
Catalog No. FAB9970U100
Add to cart
This item is not returnable. View return policy
Description
Description
Asialoglycoprotein Receptor 2 Monoclonal antibody specifically detects Asialoglycoprotein Receptor 2 in Human samples. It is validated for Flow Cytometry.
Specifications
Specifications
Asialoglycoprotein Receptor 2
2327C
Flow Cytometry 0.25-1 μg/106 cells
P07307
ASGPR 2, ASGP-R 2, ASGPR2, ASGP-R2, asialoglycoprotein receptor 2, CLEC4H2, CLEC4H2FLJ60040, C-type lectin domain family 4 member H2, HBxAg-binding protein, HBXBP, Hepatic lectin H2, HL-2
Chinese hamster ovary cell line CHO-derived recombinant human ASGR2, Gln80-Ala311, Accession No. P07307
Protein A or G purified from hybridoma culture supernatant
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied.
Monoclonal
Detects human ASGR2 in direct ELISAs.
Flow Cytometry
Alexa Fluor 350
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide.
Purified, Supernatant, Conjugated
Rabbit
IgG
100 μg
Primary
433.0
Human