Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ASIC 3 Guinea Pig anti-Rat, Polyclonal, Neuromics™ Non-distribution product as customer accommodation.

Guinea Pig Polyclonal Antibody

$245.52 - $579.50

Specifications

Antigen ASIC 3
Concentration Conc. Not Determined
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
50-210-0011 Non-distribution product as customer accommodation.
View Documents
Neuromics, IncSupplier Diversity Partner
GP14105150UL
150 μL
Each for $579.50
Add to cart
 
50-210-0012 Non-distribution product as customer accommodation.
View Documents
Neuromics, IncSupplier Diversity Partner
GP1410550UL
50 μL
Each for $245.52
Add to cart
 
Description

Description

ASIC 3 Polyclonal antibody specifically detects ASIC 3 in Rat samples. It is validated for Immunocytochemistry, Immunofluorescence, Immunohistochemistry
Specifications

Specifications

ASIC 3
Immunocytochemistry, Immunofluorescence, Immunohistochemistry
Unconjugated
Guinea Pig
Rat
O35240
286920
IgG
Primary
Antibody
Conc. Not Determined
Polyclonal
Liquid
RUO
whole serum diluted in PBS with 1% BSA and 0.05% sodium azide
Accn3; acid sensing (proton gated) ion channel 3; acid sensing ion channel 3; acid sensing ion channel subunit 3; acid-sensing (proton-gated) ion channel 3; Acid-sensing ion channel 3; Amiloride-sensitive cation channel 3; amiloride-sensitive cation channel 3, testis; ASIC3; AW742291; BOS_4613; Dorsal root ASIC; Drasic; hASIC3; hTNaC1; modulatory subunit of ASIC2a; Neuronal amiloride-sensitive cation channel 3; proton gated cation channel DRASIC; proton-gated cation channel subunit; SLNAC1; Testis sodium channel 1; TNAC1
(C)TASLDPDDFDPEPSDPLGSP
Polyclonal
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
ASIC3
SDS
Product Certifications
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit