Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ASIC Beta Guinea Pig anti-Rat, Polyclonal, Neuromics™ Non-distribution product as customer accommodation.

Guinea Pig Polyclonal Antibody

$225.68 - $501.42

Specifications

Antigen ASIC Beta
Concentration Conc. Not Determined
Applications Immunohistochemistry
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
50-209-9994 Non-distribution product as customer accommodation.
View Documents
Neuromics, IncSupplier Diversity Partner
GP10101150UL
150 μL
Each for $501.42
Add to cart
 
50-209-9995 Non-distribution product as customer accommodation.
View Documents
Neuromics, IncSupplier Diversity Partner
GP1010150UL
50 μL
Each for $225.68
Add to cart
 
Description

Description

ASIC Beta Polyclonal antibody specifically detects ASIC Beta in Rat samples. It is validated for Immunohistochemistry
Specifications

Specifications

ASIC Beta
Immunohistochemistry
Unconjugated
Guinea Pig
Rat
AGSELDEGDDSPRDLV
Polyclonal
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Conc. Not Determined
Polyclonal
Liquid
RUO
whole serum with 0.05% sodium azide
IgG
Primary
Antibody
SDS
Product Certifications
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit