Learn More
ASIC1 Polyclonal Antibody, Invitrogen™
Guinea Pig Polyclonal Antibody
Supplier:  Thermo Scientific PA124648
Catalog No. PIPA124648
Item Discontinued This item has been discontinued by the supplier and is no longer available. Please reach out to your sales representative.
Description
Description
PA1-24648 detects ASIC beta from rat samples. PA1-24648 has been successfully used in immunohistochemistry (frozen) procedures. The PA1-24648 immunogen is a synthetic peptide corresponding to residues A(3) G S E L D E G D D S P R D L V(18) of rat ASIC beta.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
ASIC1
Polyclonal
Asic1
ASIC1
Synthetic peptide corresponding to residues A(3) G S E L D E G D D S P R D L V(18) of rat ASIC beta.
RUO
79123
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Frozen)
Unconjugated
P55926
Guinea Pig
50 μL
Primary
Rat
Antibody
IgG
Recommendations
Recommendations