Learn More
ASNA1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 154501AP
Catalog No. 50-173-0238
Add to cart
Description
Description
ASNA1 is the human homolog of the bacterial arsA gene. In E. coli, ArsA ATPase is the catalytic component of a multisubunit oxyanion pump that is responsible for resistance to arsenicals and antimonials.
Specifications
Specifications
ASNA1
0.23 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O43681, O54984
ARSA, ARSA I, ARSA1, ASNA1, ATPase ASNA1, hARSA I, hASNA I, TRC40
Rabbit
IgG
150 μL
-20°C
Primary
288919, 439, 56495
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation
Unconjugated
ASNA1
Liquid
Asna1
ASNA1 Fusion Protein Ag7713
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat