Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ASPA Mouse anti-Human, Clone: OTI2D2, lyophilized, TrueMAB
Mouse Monoclonal Antibody
Manufacturer: OriGene CF805327
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
ASPA is an enzyme that catalyzes the conversion of N-acetyl_L-aspartic acid (NAA) to aspartate and acetate. NAA is abundant in the brain where hydrolysis by aspartoacylase is thought to help maintain white matter. This protein is an NAA scavenger in other tissues.Specifications
ASPA | |
Monoclonal | |
Unconjugated | |
ASPA | |
Lyophilized | |
ASPA | |
Human recombit protein fragment corresponding to amino acids 77-313 of human ASPA produced in E.coli. | |
Affinity Chromatography | |
RUO | |
Antibody | |
Monoclonal | |
Human |
Immunohistochemistry (Paraffin), Western Blot | |
OTI2D2 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
P45381 | |
ACY2, ASP | |
Mouse | |
IgG2a | |
100 μg | |
-20° C, Avoid Freeze/Thaw Cycles | |
Primary | |
443 |
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title