Learn More
BioVision IncSupplier Diversity Partner Asparaginase Activity Colorimetric/Fluorometric Assay Kit: Asparaginase Activity Assay Kit: High-Throughput Suitable Colorimetric & Fluorometric Assay for Measuring Asparaginase Activity in Tissues, Cells etc. Simple, Direct & Sensitive. 100 Assays DFS Item
Manufacturer:  BioVision IncSupplier Diversity Partner K754100
Asparaginase Activity Colorimetric/Fluorometric Assay Kit: Asparaginase (EC 3.5.1.1) is a homotetramer that catalyzes the hydrolysis of asparagine to aspartic acid and ammonia and exhibits about a 2-4% activity on glutamine and 5% on D-asparagine. Asparaginase does not occur naturally in humans but is found in bacteria, plants and many animals (e.g. guinea pigs). The enzyme has been used to reduce acrylamide, a suspected carcinogen, produced in fried starchy food products and to treat acute lymphoblastic leukemia (ALL) and some other hematopoietic neoplasms (e.g. multiple myeloma). BioVision provides a simple, direct and automation-ready procedure for measuring asparaginase activity in biological samples. In the assay, Asparaginase hydrolyzes asparagine to generate aspartic acid, which can be detected colorimetrically (?=570 nm) or fluorescently (Ex/Em = 535/590 nm) using a coupled enzymatic reaction. BioVision charges an Ice pack and Insulated Container fee of $25.00 for each order.
Catalog No. 50-111-9878