Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Asparagine synthetase Mouse anti-Human, Janelia Fluor™ 646, Clone: OTI1A10, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP270215JF646

Catalog No. N270215J646


Add to cart

Description

Description

Asparagine synthetase Monoclonal antibody specifically detects Asparagine synthetase in Human samples. It is validated for Immunocytochemistry, Immunofluorescence.
Specifications

Specifications

Asparagine synthetase
OTI1A10
Immunocytochemistry/Immunofluorescence
Affinity Purified, Conjugated
ASNS
Full length recombinant protein of human Asparagine synthetase (NP_597680) produced in HEK293T cells.
Immunogen affinity purified
Store at 4°C in the dark.
Monoclonal
Human
Immunocytochemistry, Immunofluorescence
Janelia Fluor 646
50mM Sodium Borate with 0.05% Sodium Azide
asparagine synthetase, asparagine synthetase (glutamine-hydrolyzing), asparagine synthetase [glutamine-hydrolyzing], Cell cycle control protein TS11, EC 6.3.5.4, Glutamine-dependent asparagine synthetase, TS11, TS11 cell cycle control protein
Mouse
IgG2b
0.1 mL
Primary
440.0
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit