Learn More
Aspartate Aminotransferase Mouse anti-Human, Mouse, DyLight 488, Clone: 1255, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP243716G
Catalog No. NBP243716G
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Specifications
Aspartate Aminotransferase
1255
Western Blot
Purified
GOT1
Recombinant protein encompassing a sequence within the N-terminus region of human Aspartate aminotransferase. The exact sequence is proprietary.
Protein A purified
Lipid and Metabolism
Primary
2805
Western Blot
DyLight 488
50mM Sodium Borate with 0.05% Sodium Azide
aspartate aminotransferase, cytoplasmic, EC 2.6.1.1, GIG18, Glutamate oxaloacetate transaminase 1, glutamic-oxaloacetic transaminase 1, soluble (aspartate aminotransferase 1), growth-inhibiting protein 18, Transaminase A
Mouse
IgG2b
0.1 ml
Store at 4C in the dark.
Monoclonal
Human, Mouse