Learn More
BioVision IncSupplier Diversity Partner Aspartate Colorimetric/Fluorometric Assay Kit: Colorimetric & Fluorometric Assay to Measure Aspartate in a variety of Samples such as Cell extract, Serum etc. within 40 min. Simple & Convenient. Detection Range between 0.1 – 10 nmoles. 100 Assays. DFS Item
Manufacturer:  BioVision IncSupplier Diversity Partner K552100
Aspartate Colorimetric/Fluorometric Assay Kit: L-Aspartic acid (Asp) is one of the 20 proteinogenic amino acids as building block for protein. Although a non-essential amino acid in mammals, it is a precursor to several other amino acids including four essential amino acids (Met, Thr, Ile and Lys). Aspartate is a metabolite in the urea cycle, participates in gluconeogenesis and transports reducing equivalents between the cytosol and the mitochondria via the malate-aspartate shuttle and it serves as an excitatory neurotransmitter in the brain and is an excitotoxin. BioVision's Aspartate Assay Kit provides a simple, convenient assay to measure aspartate in a variety of samples. In the assay, aspartate is converted to pyruvate which is oxidized with the conversion of a probe into a highly colored (570 nm) and fluorescent (Ex/Em 535/587 nm) species proportional to the amount of aspartate in samples. Aspartate can be quantified in the range between 0.1¿10 nmoles/well (2-200 ¿M).
Catalog No. 50-111-9766