Learn More
Abnova™ ASPHD1 293T Cell Transient Overexpression Lysate (Denatured)
Human overexpression lysate
Manufacturer:  Abnova™ H00253982T01
Catalog No. 89-935-874
Add to cart
Description
Description
Specifications
Specifications
293T
NM_181718.3
pCMV-ASPHD1 full-length
100 μL
Human
ASPHD1 293T Cell Transient Overexpression Lysate(Denatured)
Theoretical M.W. 47.2kDa
Transient overexpression cell lysate was tested with anti-ASPHD1 antibody (Mfr. No. H00253982-B01) by Western Blot.
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)
Western Blot