Learn More
Lab-Aids™ Aspirin Study Kit Products for Science Education
Relate science to the real world, calculate the percent binder in tablets as well as its composition, determine pH of tablet solution and calculate the unit cost as well as cost-benefits.
Manufacturer:  Lab-Aids™ 180
Catalog No. S19470
Add to cart
Includes: Brand “X” aspirin, sodium carbonate solution, iron nitrate solution, Benedict's solution, plastic dropper, and Lugol's solution
Universal indicator paper and blue litmus paper
Sample stock bottles
30 test tubes
30 student worksheets and guides
Instruction manual with MSDS
This item is not returnable. View return policy
Description
Description
Aspirin (acetylsalicylic acid) is probably the most used and abused non-prescription drug. Using aspirin tablets brought from home, students analyze and compare them with the Brand 'X' aspirin included in this Lab-Aid. They determine the percentage of aspirin and 'filler' components in a tablet, check pH and calculate the per aspirin cost.
Students will draw conclusions concerning which aspirins are better, which are safer, and which do nothing at all. This kit makes science more interesting because it brings 'real world' questions into the classroom.
Extension activities are possible with this kit. Note: Teacher and/or students must supply additional various brands of aspirin. Accommodates two classes, each with 15 groups of 2 students. Includes Teacher guide, student sheets, assessment questions, SDS for all chemicals in kit.
Specifications
Specifications
Teacher guide with MSDS, 30 student worksheets and guides, 30 student data sheets, Lugol's solution, bottle, brand X aspirin, 1 bottle, 1 dropper, 5 labeled sample stock bottles, 30 test tubes, 1 vial of blue litmus paper, 1 vial of universal indicator paper, 2 drop control bottles of Benedict's solution, 2 drop control bottles of Iron Nitrate solution, 2 drop control bottles of Sodium Carbonate solution
To calculate the percent binder in tablets as well as its composition, determine ph of tablet solution
30 students
Aspirin Study Kit