Learn More
Abnova™ ASPN 293T Cell Transient Overexpression Lysate (Denatured)
Human overexpression lysate
Manufacturer:  Abnova™ H00054829T02
Catalog No. 89-935-875
Add to cart
Description
Description
Specifications
Specifications
293T
BC063114
pCMV-ASPN full-length
100 μL
Human
ASPN 293T Cell Transient Overexpression Lysate(Denatured)
Theoretical M.W. 43.9kDa
Transient overexpression cell lysate was tested with anti-ASPN antibody (Mfr. No. H00054829-D01P) by Western Blot.
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)
Western Blot