Learn More
Abnova™ ASPSCR1 293T Cell Transient Overexpression Lysate (Denatured) (T02)
Human overexpression lysate
Manufacturer:  Abnova™ H00079058T02
Catalog No. 89-935-877
Add to cart
Description
Description
Specifications
Specifications
293T
BC018722.1
pCMV-ASPSCR1 full-length
100 μL
Human
ASPSCR1 293T Cell Transient Overexpression Lysate(Denatured)
Theoretical M.W. 60.2kDa
Transient overexpression cell lysate was tested with anti-ASPSCR1 antibody (Mfr. No. H00079058-D01P) by Western Blot.
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)
Western Blot