Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ASRGL1 Mouse anti-Human, Allophycocyanin/Cy7, Clone: CRASH/1289, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP254450APCCY7

Catalog No. N254450APC7


Add to cart

Description

Description

ASRGL1 Monoclonal antibody specifically detects ASRGL1 in Human samples. It is validated for Flow Cytometry.
Specifications

Specifications

ASRGL1
CRASH/1289
Flow Cytometry
Conjugated, Purified
ASRGL1
Recombinant human full-length ASRGL1
Protein A or G purified
Store at 4°C in the dark. Do not freeze.
Monoclonal
Human
Flow Cytometry
APC-Cyanine7
PBS with 0.05% Sodium Azide
ALP1, ALPL-asparagine amidohydrolase, asparaginase like 1, asparaginase-like 1 protein, Asparaginase-like protein 1, CRASH, EC 3.5.1.1, FLJ22316, L-asparaginase
Mouse
IgG1
0.1 mL
Primary
80150.0
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit