Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ASRGL1 Mouse anti-Human, Clone: CRASH/1290, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP30736810UG

 View more versions of this product

Catalog No. NB122209


Add to cart

Description

Description

ASRGL1 Monoclonal specifically detects ASRGL1 in Human samples. It is validated for ELISA.
Specifications

Specifications

ASRGL1
CRASH/1290
Unconjugated
10 mM PBS with 0.05% BSA
ALP1, ALPL-asparagine amidohydrolase, asparaginase like 1, asparaginase-like 1 protein, Asparaginase-like protein 1, CRASH, EC 3.5.1.1, FLJ22316, L-asparaginase
Recombinant human ASRGL1 fragment
Protein A or G purified
RUO
Primary
80150
ELISA
0.2 mg/ml
ELISA
Purified
Mouse
IgG1 Kappa
100 μg
Store at 4°C.
Monoclonal
Human
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit