Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ASRGL1 Mouse anti-Human, Clone: CRASH/1290, Novus Biologicals™

Mouse Monoclonal Antibody

$138.37 - $352.89

Specifications

Antigen ASRGL1
Clone CRASH/1290
Concentration 0.2 mg/ml
Dilution ELISA
Applications ELISA
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB122210
View Documents
Novus Biologicals
NBP30736820UG
20 μg
Each for $138.37
Add to cart
 
NB122209
View Documents
Novus Biologicals
NBP30736810UG
100 μg
Each for $352.89
Add to cart
 
Description

Description

ASRGL1 Monoclonal antibody specifically detects ASRGL1 in Human samples. It is validated for ELISA
Specifications

Specifications

ASRGL1
0.2 mg/ml
ELISA
Purified
RUO
10 mM PBS with 0.05% BSA
80150
IgG1 Kappa
Primary
Store at 4°C.
CRASH/1290
ELISA
Unconjugated
Mouse
Human
ALP1, ALPL-asparagine amidohydrolase, asparaginase like 1, asparaginase-like 1 protein, Asparaginase-like protein 1, CRASH, EC 3.5.1.1, FLJ22316, L-asparaginase
Recombinant human ASRGL1 fragment
Monoclonal
Protein A or G purified
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit