Learn More
ASS1 Mouse anti-Human, Clone: OTI8C4, lyophilized, TrueMAB™ Non-distribution product as customer accommodation.
Mouse Monoclonal Antibody
Manufacturer:  OriGeneSupplier Diversity Partner CF809122
Catalog No. 50-166-9285
Add to cart
Description
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
The protein encoded by this gene catalyzes the penultimate step of the arginine biosynthetic pathway. There are approximately 10 to 14 copies of this gene including the pseudogenes scattered across the human genome, among which the one located on chromosome 9 appears to be the only functional gene for argininosuccinate synthetase. Mutations in the chromosome 9 copy of ASS cause citrullinemia. Two transcript variants encoding the same protein have been found for this gene.
Specifications
Specifications
ASS1
Monoclonal
Unconjugated
ASS1
Lyophilized
ASS1
Synthetic peptide corresponding to residues near C-terminus of human ASS1.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Western Blot
OTI8C4
PBS with 8% trehalose and no preservative; pH 7.3
P00966
ASS, CTLN1
Mouse
IgG1
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
445