Learn More
Thermo Scientific™ Assembled Caps and Septa for 8-425 Standard-Opening Vials Available on GSA/VA Contract for Federal Government customers only.
Thermo Scientific™ National Assembled Caps and Septa for 8-425 Standard-Opening Vials are available in a variety of colors and septum materials.
Manufacturer:  Thermo Scientific™ C401361Y
 View more versions of this product
Catalog No. 03-378-356
  Check Availability
Add to cart
Description
Description
Thermo Scientific™ National Assembled Caps and Septa for 8-425 Standard-Opening Vials are available in a variety of colors and septum materials for a wide range of applications.
  • Open top caps can be used with any 8mm septa
  • Phenolic caps are suitable for autoclaving and low temperature applications
  • Polypropylene caps are chemically inert and suitable for most chromatography applications
  • Kim-Lok™ cap/septum assemblies feature a molded centering ring to eliminate septum movement during injection
  • PTFE/red rubber septa are economical for many GC applications
  • PTFE/silicone septa are most popular for HPLC and IC applications
  • Pre-slit septa are easier to pierce with thin-gauge needles
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
PP, Yellow; Kim-Lok Red PTFE/White Silicone septum
Polypropylene
8-425mm
8-425 Standard-Opening Vials
Yellow
Kim-Lok Red PTFE/White Silicone
Kim-Lok cap w/flange
C4013-61Y