Associated Bag Asbestos Hazard Bags

Collect, transport, and dispose of asbestos-contaminated materials easily and safely

Manufacturer: Associated Bag Company 658103C

 View more versions of this product

Catalog No. 19-821-943

  Check Availability
Add to Cart

    Specifications

    Product Type Asbestos Hazard Bag
    Certifications/Compliance OSHA,1910.1001
    For Use With (Application) Collect, transport and dispose of asbestos contaminated materials
    Height (English) 37 in.
    Includes Biohazard label
    Thickness (English) 6mil
    Thickness (Metric) 0.152mm
    Color Clear
    Length (English) 30 in.

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit