Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIP™ Assurance™ 18 in. Chemical Resistance Nitrile Gloves, Sandpatch Grip

Unsupported, unlined 22 mil nitrile gloves are resistant to physical and chemical hazards such as cut, puncture, solvents and oil based chemicals.

$127.00

Specifications

ANSI Abrasion Rating ANSI 4
Certifications/Compliance 21 CFR 170-199
Thickness (English) 22 mil
Thickness (Metric) 0.55 mm
Length (English) 18 in.
View More Specs

Products 4
Catalog Number Mfr. No. Size Price Quantity & Availability  
Catalog Number Mfr. No. Size Price Quantity & Availability  
19-152-500
View Documents
PIP™
50N2272G/M
Medium
Dozen of 12 for $127.00
Add to cart
 
19-152-501
View Documents
PIP™
50N2272G/L
Large
Dozen of 12 for $127.00
Add to cart
 
19-152-502
View Documents
PIP™
50N2272G/XL
X-Large
Dozen of 12 for $127.00
Add to cart
 
19-152-503
View Documents
PIP™
50N2272G/XXL
2X-Large
Dozen of 12 for $127.00
Add to cart
 
Description

Description

  • Made from component materials that comply with Federal Regulations for food contact
  • Unlined, chlorinated
  • Straight cuff
  • Color: green
  • Sandpatch grip
  • Overall Length: 18 in. (46 cm)
  • Ideal for:
    • Chemical processing
    • Oil refining and petrochemicals
    • Food processing
    • Aerospace and automotive degreasing
    • Automotive assembly and painting
Specifications

Specifications

ANSI 4
22 mil
18 in.
Sandpatch
Straight
21 CFR 170-199
0.55 mm
45.7 cm
Unlined
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit