Learn More
PIP™ Assurance™ Blue 15 mil Flock-Lined Chemical Resistant Nitrile Gloves
Unsupported lined gloves provide protection from chemical and physical hazards. Commonly used in light chemical applications, printing and meat processing.
$25.00
Specifications
Certifications/Compliance 21 CFR 170-199
Thickness (English) 15 mil
Thickness (Metric) 0.38 mm
Length (English) 13 in.
Length (Metric) 33 cm
View More Specs
Products 4
Catalog Number Mfr. No. Size Price Quantity & Availability  
Catalog Number Mfr. No. Size Price Quantity & Availability  
19-161-848
View Documents
PIP™
50N160B/S
Small
Dozen of 12 for $25.00
Case of 12 DZ for $300.00
Add to cart
19-152-504
View Documents
PIP™
50N160B/M
Medium
Dozen of 12 for $25.00
Case of 12 DZ for $300.00
Add to cart
19-161-850
View Documents
PIP™
50N160B/L
Large
Dozen of 12 for $25.00
Case of 12 DZ for $300.00
Add to cart
19-152-505
View Documents
PIP™
50N160B/XXL
2X-Large
Dozen of 12 for $25.00
Case of 12 DZ for $300.00
Add to cart
Description
Description
  • Resistant to cut, puncture, snag and abrasion
  • Resistant to many chemical hazards, including solvents and oil-based chemicals
  • Lightweight construction and hand specific design provide comfort with protection
  • Provides flexibility, tactile sensitivity and dexterity sometimes lacking in heavier gauge nitriles
  • Raised diamond grip pattern
  • Chlorinated
  • Straight cuff, Length: 13 in. (33 cm)
  • Thickness: 15 mil
  • Blue
Specifications
Specifications
21 CFR 170-199
0.38 mm
33 cm
Lined
Straight
15 mil
13 in.
Raised diamond
Cotton flock