Learn More
PIP™ Assurance™ Unlined Nitrile Gloves
Used in light chemical applications, printing, meat and poultry processing.
Manufacturer:  PIP™ 50N115G/XXL
 View more versions of this product
Catalog No. 19-121-187E
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
This item is not returnable. View return policy
Description
Description
  • Light weight construction and hand specific design provide real comfort with protection
  • Provides flexibility, tactile sensitivity and dexterity sometimes lacking in heavier gauge unsupported nitriles
  • Provides the protection from chemical and physical hazards not always found in thinner gauge nitrile disposables
  • Unlined to reduce potential of product contamination
  • Straight cuff adds additional length to protect the forearm from liquid run-off
  • Raised Diamond pattern for an enhanced grip
  • Chlorinated to harden and cleanse the surface of the glove
Specifications
Specifications
ISO 9001:2000
Green
2X-Large
0.27 mm
13 in.
No
Nitrile
11 mil
Straight
33 cm