Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PIP™ Assurance™ Unlined Nitrile Gloves
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
Description
- Light weight construction and hand specific design provide real comfort with protection
- Provides flexibility, tactile sensitivity and dexterity sometimes lacking in heavier gauge unsupported nitriles
- Provides the protection from chemical and physical hazards not always found in thinner gauge nitrile disposables
- Unlined to reduce potential of product contamination
- Straight cuff adds additional length to protect the forearm from liquid run-off
- Raised Diamond pattern for an enhanced grip
- Chlorinated to harden and cleanse the surface of the glove
Specifications
Certifications/Compliance | ISO 9001:2000 |
Chlorinated | No |
Color | Green |
Material | Nitrile |
Size | Medium |
Thickness (English) | 11 mil |
Thickness (Metric) | 0.27 mm |
For Use With (Application) | light chemical applications, printing, meat and poultry processing |
Glove Cuff Style | Straight |
Length (English) | 13 in. |
Show More |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction