Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medicom™ AssureTouch™ Pure™ Nitrile Medical Examination Gloves with Textured Fingertips

Catalog No. 17100908 Shop All Medicom Products
Change view
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-100-908 Medium
17-101-078 X-Small
17-101-079 Small
17-101-081 Large
17-101-082 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 17-100-908 Supplier Medicom™ Supplier No. 9970C
Only null left
Add to Cart
Add to Cart

AssureTouch™ Pure™ Nitrile Medical Examination Gloves with Textured Fingertips. Ideal for procedures that require a high level of precision and tactile sensitivity. DermaSense technology helps reduce the risk of skin reactions. Textured fingertips ensure a secure grip. Chemo-tested.

  • These chemo-tested, ultra-thin nitrile gloves are ideal for procedures that require precision and tactile sensitivity.
  • Textured fingertips ensure a secure grip, even when wet.
  • DermaSense technology eliminates the chemical accelerators commonly found in nitrile gloves to reduce the risk of skin reactions.

Specifications

Chlorinated No
Accelerator Free Yes
Applications Healthcare, Industrial, Laboratory
AQL 1.5
Color Blue
Disposable Single-use
Exterior Finish Fingertip Textured
Latex Free Yes
Powdered Powder Free
Size Medium
Sterility Non-sterile
FDA 510 (k) Yes
Glove Cuff Style Beaded
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.