Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ AST/GOT Reagent

Determine quantity of aspartate aminotransferase in serum or plasma in vitro with Thermo Scientific™ AST/GOT, featuring a 30-day reconstituted stability.

$141.60

Specifications

CE Marker Yes
Linearity 0 to 450U/L
Methods Compatibility Kinetic UV
Storage Requirements 2°C to 8°C
View More Specs

Products 1
Catalog Number Description Quantity Price Quantity & Availability  
Catalog Number Description Quantity Price Quantity & Availability  
TR70121
Accessories View Documents
Infinity AST/GOT 2 x 125mL
Pack of 2 for $141.60
Add to cart
 
Description

Description

Determine quantity of aspartate aminotransferase in serum or plasma in vitro with Thermo Scientific™ AST/GOT, featuring a 30-day reconstituted stability.

AST is widely distributed with high concentrations in the heart, liver, skeletal muscle, and kidney. Damage or disease to any of these tissues such as myocardial infarction, viral hepatitis, liver necrosis, cirrhosis and muscular dystrophy may result in raised serum levels of AST.

  • Reagent is suitable for manual use or on automated clinical chemistry analyzers
  • Applications for a wide range of automated analyzers are available on request.
Specifications

Specifications

Yes
0 to 450U/L
Kinetic UV
2°C to 8°C
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit