Learn More
Thermo Scientific AST/GOT Reagent
Determine quantity of aspartate aminotransferase in serum or plasma in vitro with Thermo Scientific AST/GOT, featuring a 30-day reconstituted stability.
Description
Determine quantity of aspartate aminotransferase in serum or plasma in vitro with Thermo Scientific AST/GOT, featuring a 30-day reconstituted stability.
AST is widely distributed with high concentrations in the heart, liver, skeletal muscle, and kidney. Damage or disease to any of these tissues such as myocardial infarction, viral hepatitis, liver necrosis, cirrhosis and muscular dystrophy may result in raised serum levels of AST.
- Reagent is suitable for manual use or on automated clinical chemistry analyzers
- Applications for a wide range of automated analyzers are available on request.
Specifications
CE Marked | |
Plasma, Serum | |
Reagent | |
Infinity AST/GOT | |
0 to 450U/L | |
No | |
Stable Liquid |
Liquid | |
2°C to 8°C | |
Yes | |
AST/GOT | |
Kinetic UV | |
2 x 125 mL | |
Liquid |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.