Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Astemizole, BP, Spectrum™

Click to view available options
Quantity:
5 kg
Packaging:
Poly Pail

A3456, 68844-77-9, C28H31FN4O

Spectrum™ Astemizole, BP was withdrawn from the U.S. market in 1999. Astemizole is an antihistamine used to prevent sneezing, runny nose, itching and watering of the eyes, and other allergic symptoms. Spectrum Chemical BP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Packaging Poly Pail
Quantity 5 kg
CAS 68844-77-9
CAS Max % 1
Molecular Formula C28H31FN4O
InChI Key GXDALQBWZGODGZ-UHFFFAOYSA-N
SMILES COC1=CC=C(CCN2CCC(CC2)NC2=NC3=CC=CC=C3N2CC2=CC=C(F)C=C2)C=C1
IUPAC Name 1-[(4-fluorophenyl)methyl]-N-{1-[2-(4-methoxyphenyl)ethyl]piperidin-4-yl}-1H-1,3-benzodiazol-2-amine
Molecular Weight (g/mol) 458.58