Learn More
ASTM™ D6584 1,2,4-Butanetriol solution, Honeywell Fluka™
1000 μg/mL in pyridine, analytical standard, reference material
Manufacturer:  Honeywell Chemicals R13845ML
Catalog No. 60-049-103
Add to cart
Description
Description
Ensure you have the information you need to quickly start your analysis, and pass relevant audits:
  • High quality inorganic and organic reference materials produced according to ISO 9001
  • Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results
  • Free expert technical support
Specifications
Specifications
110-86-1, 42890-76-6
Analytical Standard
0.98 g/cm3
5 mL
BIOFUELS
Liquid
Glass Bottle
115°C
20°C
Safety and Handling
Safety and Handling
P284+P210+P243+P260+P280
H225+H302+H312+H332
Danger