Learn More
ASTM™ D6584 Standard Solution 1, Certified Reference Material, in Pyridine (varied conc.), MilliporeSigma™ Supelco™
This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.
Manufacturer:  MilliporeSigma Supelco CRM44899
Catalog No. 11-101-6442
Add to cart
Description
Description
Specifications
Specifications
TraceCERT™ Analytical Standard
1,3-Diolein 50 µg/mL; Glycerol 5 µg/mL; Monoolein 100 µg/mL; Triolein 50 µg/mL
Ampule
1 mL
SDS
Documents
Product Certifications
Promotions
Promotions
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
ASTM is a registered trademark of American Society for Testing and Materials