Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ASTN2 Rabbit anti-Mouse, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA549239
Description
This gene encodes a protein that is expressed in the brain and may function in neuronal migration, based on functional studies of the related astrotactin 1 gene in human and mouse. A deletion at this locus has been associated with schizophrenia. Multiple transcript variants encoding different proteins have been found for this locus. [provided by RefSeq, May 2010]Specifications
ASTN2 | |
Polyclonal | |
PBS with 0.09% sodium azide | |
Q80Z10 | |
ASTN2 | |
KLH conjugated synthetic peptide between 220-248 amino acids from the N-terminal region of human ASTN2. | |
400 μL | |
Primary | |
Mouse | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
Astn2 | |
Astrotactin-2, ASTN2, KIAA0634 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
56079 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction