Learn More
Astral Diagnostics™ NEAT STAIN™ Staining Systems for Hematology, Microbiology and Parasitology
Offer fast, clean, convenient and cost-effective staining while achieving consistent, reproducible results
Manufacturer:  Astral Diagnostics™ NSH5
 View more versions of this product
Catalog No. 22-246488
Add to cart
Description
Description
  • All procedures are well-defined and consist of simple dip steps
  • Each ready-to-use, pre-filled staining cuvet performs a minimum of 25 tests
  • Each shelf pack contains six cuvets, a holding tray, reclosures and complete instructions
  • An easy-to-use, optional, five-slide holder is available
Specifications
Specifications
5 Slide Holder
Each
Staining for microbiology
NEAT Stain
Documents
Accessories