Learn More
Astral Diagnostics™ Quick III™ Set
A three-step rapid differential stain for the differential staining of blood smears, blood parasites, microbiological organisms, and bone marrow specimens
Manufacturer:  Astral Diagnostics™ 5316
Catalog No. 22-246-493
Add to cart
Description
Description
  • Has extensive applicability in cytology for use in fine needle aspiration analysis
  • Processes specimens in 30 seconds and utilizes a fixative and two staining solutions to achieve the desired results
  • Staining intensity can be adjusted to suit the requirements of the user by adjusting the times in one or both staining solutions
Specifications
Specifications
Kit
A three step rapid differential stain for the differential staining of Blood smears, Blood parasites, microbiological organisms and bone marrow specimens, has extensive applicability in cytology for use in fine needle aspiration analysis, can also be used as an andrology stain