Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Astrin Antibody, HRP, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB500132H
Description
Astrin Polyclonal antibody specifically detects Astrin in Human samples. It is validated for Western BlotSpecifications
Astrin | |
Polyclonal | |
PBS | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot | |
HRP | |
Astrin, Deepest, hMAP126, MAP126mitotic spindle coiled-coil related protein, mitotic spindle associated protein p126, Mitotic spindle-associated protein p126, sperm associated antigen 5, sperm-associated antigen 5 | |
MAP126 protein | |
0.1 mL | |
Cell Cycle and Replication, Chromatin Research, Core ESC Like Genes, MAP Kinase Signaling, Stem Cell Markers | |
10615 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Product Content Correction